Skip to Content
Merck
All Photos(1)

Key Documents

930873

Sigma-Aldrich

[3-(2,4,6-Trimethylbenzyl)-1-(2,6-diisopropylphenyl)-4,5-dimethylimidazolylidene]copper chloride

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H36ClCuN2
Molecular Weight:
487.59
UNSPSC Code:
12352101
NACRES:
NA.22

Quality Level

Assay

≥95%

form

powder

Application

[3-(2,4,6-Trimethylbenzyl)-1-(2,6-diisopropylphenyl)-4,5-dimethylimidazolylidene]copper chloride is an NHC catalyst bearing a highly hindered and flexible unsymmetrical NHC ligand for hydrofunctionalization reactions. This catalyst can be used in hydroelement additions, element-element additions, carbo-element additions, hydrations, hydroaminations, hydroarylations, hyroalkoxylations, hydrocarboxylations, cyclizations, hydrofluorinations, as well as a variety of combined processes using tandem reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service