Skip to Content
Merck
All Photos(3)

Key Documents

929719

Sigma-Aldrich

ElectroGreen®

greener alternative

NMP substitute for electronics - type 1, bio-sourced

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000
NACRES:
NA.05

vapor pressure

~1.0 mmHg ( 20 °C)

Quality Level

description

Relative Evaporation rate: <0.05
Hansen solubility parameters: SPd 8.8; SPp 5.9; SPh 3.7

for electronic purposes

Assay

≥99% (GC)

form

liquid

greener alternative product characteristics

Design for Energy Efficiency
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.

impurities

≤1 ppm As, Cr, Cd, Cu, Hg, Mn, Ni, Pb, trace (each)
≤3 ppm Zn, trace
0.03 wt. % Acidity (as lactic acid)
0.04 wt. % Water

evapn. residue

≤0.05%

color

clear

viscosity

16.1  cP(20 °C)

bp

~154 °C (Initial)

density

1.094 at 25 °C (Specific gravity)

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

This ElectroGreen® solvent is made of bio-based(corn/beets-derived) safer alternatives to replace synthetic NMP fossil-based solvent with specifications tailored for electronics and energy applications. All our ElectroGreen® solvents line are derived from a renewable resource (verified through C14 ASTM D6866-16testing). We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Click here for more details.

This solvent blend consists of propylene carbonate 50-70%, Ethyl lactate 30-50%; ethyl 3-ethoxypropionate 20-30%.

Application

This is our bio-based replacement for N-Methyl Pyrrolidone (NMP), a highly used solvent in electronics applications. It has low acidity and water contents, with moderate drying time, and no film residue. It has a flash point over 60 ºC, with excellent solvency for coating and ink formulations. Low vapor pressure and high loading capacity. It is easy and inexpensive to distill/recycle, with no environmentally hazardous ingredients and no Hazardous Air Pollutants.

Storage and Stability

To ensure low water content, handle under inert conditions. Product should be refrigerated upon receipt.

Legal Information

ElectroGreen is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

156.9 °F

Flash Point(C)

69.4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Carbon-based Sustainable Organic Electronics (SOE) limit the use of critical elements and biodegrade at their end-of-life. This review offers insight on how structural and energy disorder in these materials influence device performance and includes evaluations of various transport models and their limitations.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service