Skip to Content
Merck
All Photos(1)

Key Documents

906506

Sigma-Aldrich

HLAM-AβIδ1MεLVproSTγ-13CH3 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

technique(s)

bio NMR: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

General description

HLAM-AβIδ1MεLVproSTγ-13CH3 kit has 13CH3 isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

For detection of long-range nOes or for the NMR study of large proteins
HLAM-AβIδ1MεLVproSTγ-13CH3 is an optimized kit used to produce proteins with 13CH3 isotopomer on methyl groups of alanine, isoleucine, methionine, leucine and threonine residues. This kit has been tested with protein isotopic labeling in E. coli. It can be used to study large molecular system and complex protein structure in NMR spectroscopy along with other spectroscopic approaches. The sensitivity of the isotopomer can be enhanced by three-fold multiplicity. This kit can also be used to simplify crowded spectra to provide specific probes of dynamic and structural studies in solid state NMR spectroscopy.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service. jj

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

César Fernández et al.
Current opinion in structural biology, 13(5), 570-580 (2003-10-22)
Transverse relaxation-optimized spectroscopy (TROSY), in combination with various isotope-labeling techniques, has opened avenues to study biomolecules with molecular masses of up to 1000000Da by solution NMR. Important recent applications of TROSY include the structure determination of membrane proteins in detergent
Silke Wiesner et al.
Current opinion in structural biology, 35, 60-67 (2015-09-26)
Intermolecular interactions are indispensible for biological function. Here we discuss how novel NMR techniques can provide unique insights into the assembly, dynamics and regulation of biomolecular complexes. We focus on applications that exploit the methyl TROSY effect and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service