742872
1-Ethyl-3-methylimidazolium chloride-aluminum chloride
Synonym(s):
AICl3-EMIC, Aluminum chloride-1-Ethyl-3-methylimidazolium chloride, EMIM Cl AlCl3
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C6H11ClN2)2 · (AlCl3)3
Molecular Weight:
693.26
UNSPSC Code:
12352100
NACRES:
NA.22
Recommended Products
Related Categories
Application
1-Ethyl-3-methylimidazolium chloride-aluminum chloride can be used as a medium for the preparation of poly(p-phenylene) films via electropolymerization of benzene.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Benzene polymerization in 1-ethyl-3-methylimidazolium chloride-AlCl3 ionic liquid.
Goldenberg LM and Osteryoung RA.
Synt. Metals, 64(1), 63-68 (1994)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service