Skip to Content
Merck
All Photos(1)

Key Documents

727490

Sigma-Aldrich

Azidobenzene solution

~0.5 M in tert-butyl methyl ether

Synonym(s):

Phenyl azide solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
Beilstein:
742248
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

≥95.0% (HPLC)

Quality Level

form

liquid

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

functional group

azide

storage temp.

−20°C

SMILES string

[N-]=[N+]=Nc1ccccc1

InChI

1S/C6H5N3/c7-9-8-6-4-2-1-3-5-6/h1-5H

InChI key

CTRLRINCMYICJO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F

Flash Point(C)

4 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Laëtitia Renaudie et al.
Biomacromolecules, 8(2), 679-685 (2007-02-13)
Grafting of a new carbohydrate UV-reactive molecule, an azidophenyl lactamine (AzPhLac), was achieved on fibers of three different diameters: 12, 18, and 32 microm. Adsorption of AzPhLac on fibers was obtained by using the dip-coating method in solution. The effect
C Malaveille et al.
Cancer research, 42(4), 1446-1453 (1982-04-01)
3-Methyl-1-phenyltriazene and a series of ring-substituted derivatives (4-methylphenyl, 4-chlorophenyl, and 2,4,6-trichlorophenyl), structurally related benzenediazonium fluoborates and phenyl azides, as well as the recently isolated [1-methyl-3-(2,4,6-trichlorophenyl)-2-triazeno]methyl-beta-D-glucopyranoside uronic acid, were studied for their mutagenic activity in Salmonella typhimurium strains. Of these compounds
Pascal Viel et al.
Langmuir : the ACS journal of surfaces and colloids, 29(6), 2075-2082 (2013-01-16)
Covalent immobilization of unmodified biological materials as proteins has been performed through a one-step and soft method. This process is based on a polyazidophenylene layer derived from the electroreduction of the parent salt 4-azidobenzenediazonium tetrafluoborate on gold substrates. The wavelength
Jacob Gubbens et al.
Chemistry & biology, 16(1), 3-14 (2009-01-28)
New lipid analogs mimicking the abundant membrane phospholipid phosphatidylcholine were developed to photocrosslink proteins interacting with phospholipid headgroups at the membrane interface. In addition to either a phenylazide or benzophenone photoactivatable moiety attached to the headgroup, the lipid analogs contained
M Zofall et al.
Nucleic acids research, 28(21), 4382-4390 (2000-11-01)
Two new photoreactive dATP analogs, N(6)-[4-azidobenzoyl-(2-aminoethyl)]-2'-deoxyadenosine-5'-triphospha+ ++ te (AB-dATP) and N(6)-[4-[3-(trifluoromethyl)-diazirin-3-yl]benzoyl-(2-aminoethyl) ]-2 '-deoxyadenosine-5'-triphosphate (DB-dATP), were synthesized from 2'-deoxyadenosine-5'-monophosphate in a six step procedure. Synthesis starts with aminoethylation of dAMP and continues with rearrangement of N(1)-(2-aminoethyl)-2'-deoxyadenosine-5'-monophosphate to N(6)-(2-aminoethyl)-2'-deoxyadenosine-5'-monophosphate (N(6)-dAMP). Next, N(6)-dAMP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service