Skip to Content
Merck
All Photos(1)

Key Documents

706434

Sigma-Aldrich

(S)-N-Fmoc-piperidine-2-carboxylic acid

97%

Synonym(s):

Fmoc-Pip-OH, (S)-1-Fmoc-piperidine-2-carboxylic acid, N-Fmoc-L-pipecolinic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H21NO4
CAS Number:
Molecular Weight:
351.40
Beilstein:
4718405
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.26

Assay

97%

form

powder

optical activity

[α]22/D −23°, c = 1 in DMF

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

153-158 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H]1CCCCN1C(=O)OCC2c3ccccc3-c4ccccc24

InChI

1S/C21H21NO4/c23-20(24)19-11-5-6-12-22(19)21(25)26-13-18-16-9-3-1-7-14(16)15-8-2-4-10-17(15)18/h1-4,7-10,18-19H,5-6,11-13H2,(H,23,24)/t19-/m0/s1

InChI key

CKLAZLINARHOTG-IBGZPJMESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service