Skip to Content
Merck
All Photos(1)

Key Documents

703346

Sigma-Aldrich

Iodomethane-13C solution

NMR reference standard, 1% in chloroform-d ("100%", 99.96 atom % D), chromium (III) acetylacetonate 0.2 %, trimethyl phosphite 1 %, 99 atom % 13C, NMR tube size 6.5 mm × 8 in.

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CH3I
Molecular Weight:
142.93
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

grade

NMR reference standard

Quality Level

isotopic purity

99 atom % 13C

form

solution

composition

chromium (III) acetylacetonate, 0.2%
trimethyl phosphite, 1%

concentration

1% in chloroform-d ("100%", 99.96 atom % D)

technique(s)

NMR: suitable

NMR tube size

6.5 mm × 8 in.

mass shift

M+1

SMILES string

[13CH3]I

InChI

1S/CH3I/c1-2/h1H3/i1+1

InChI key

INQOMBQAUSQDDS-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Indirect detection test

Quantity

6.5 mm O.D. tube contains 1.2 mL.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Oral - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service