Skip to Content
Merck
All Photos(1)

Key Documents

700665

Sigma-Aldrich

(11bR)-2,6-Di-9-phenanthrenyl-4-hydroxy-dinaphtho[2,1-d:1′,2′-f][1,3,2]dioxaphosphepin-4-oxide

Synonym(s):

(R)-3,3′-Bis(9-phenanthryl)-1,1′-binaphthalene-2,2′-diyl hydrogen phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H29O4P
CAS Number:
Molecular Weight:
700.72
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

optical activity

[α]22/D −44.0°, c = 1 in DMSO

mp

390-400 °C

storage temp.

−20°C

SMILES string

OP1(=O)Oc2c(cc3ccccc3c2-c4c(O1)c(cc5ccccc45)-c6cc7ccccc7c8ccccc68)-c9cc%10ccccc%10c%11ccccc9%11

InChI

1S/C48H29O4P/c49-53(50)51-47-43(41-25-29-13-1-5-17-33(29)37-21-9-11-23-39(37)41)27-31-15-3-7-19-35(31)45(47)46-36-20-8-4-16-32(36)28-44(48(46)52-53)42-26-30-14-2-6-18-34(30)38-22-10-12-24-40(38)42/h1-28H,(H,49,50)

InChI key

BVICVEIKBNVROI-UHFFFAOYSA-N

Application

Catalyst for:
  • Asymmetric hydrogenation cascade
  • Enantioselective Friedel-Crafts reaction
  • Asymmetric hydrogenation of 2-substituted quinolines
  • Chiral Broensted acid catalyzed enantioselective a-aminoxylation of ene carbamates
  • Preparation of β-carbolines via diastereo- and enantioselective N-acyliminium cyclization cascades of tryptamines and keto acids/esters
  • Asymmetric transfer hydrogenation of substituted quinoxalines

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service