Skip to Content
Merck
All Photos(1)

Key Documents

684864

Sigma-Aldrich

2,4-Dichloro-5-(trifluoromethyl)pyrimidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5HCl2F3N2
CAS Number:
Molecular Weight:
216.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.475

density

1.609 g/mL at 25 °C

functional group

chloro
fluoro

storage temp.

2-8°C

SMILES string

FC(F)(F)c1cnc(Cl)nc1Cl

InChI

1S/C5HCl2F3N2/c6-3-2(5(8,9)10)1-11-4(7)12-3/h1H

InChI key

IDRUEHMBFUJKAK-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Eye Irrit. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

199.9 °F

Flash Point(C)

93.3 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service