657131
Bis(4-bromophenyl)amine
97%
Synonym(s):
4,4′-Dibromodiphenylamine, N-phenyl-4,4′-dibromoaniline
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H9Br2N
CAS Number:
Molecular Weight:
327.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
form
solid
mp
105-108 °C
functional group
bromo
SMILES string
Brc1ccc(Nc2ccc(Br)cc2)cc1
InChI
1S/C12H9Br2N/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8,15H
InChI key
VKVHTZNHLOGHGP-UHFFFAOYSA-N
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4 - Eye Dam. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service