Skip to Content
Merck
All Photos(1)

Documents

638587

Sigma-Aldrich

3-Methylbenzothiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8S
CAS Number:
Molecular Weight:
148.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

refractive index

n20/D 1.6240 (lit.)

bp

72-74 °C/2 mmHg (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

Cc1csc2ccccc12

InChI

1S/C9H8S/c1-7-6-10-9-5-3-2-4-8(7)9/h2-6H,1H3

InChI key

SEBRPHZZSLCDRQ-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

219.2 °F - closed cup

Flash Point(C)

104 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yujuan Hua et al.
Journal of separation science, 41(2), 469-474 (2017-11-11)
A fast and reliable analytical technique for the determination of total sulfur levels in complex hydrocarbon matrices is introduced. The method employed flow injection technique using a gas chromatograph as a sample introduction device and a gas phase dual-plasma sulfur

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service