About This Item
Recommended Products
isotopic purity
≥98 atom % D
Assay
≥99% (CP)
99% (CP)
form
solid
optical activity
[α]25/D -33.0°, c = 1 in H2O
greener alternative product score
old score: 47
new score: 32
Find out more about DOZN™ Scoring
greener alternative product characteristics
Less Hazardous Chemical Syntheses
Safer Solvents and Auxiliaries
Use of Renewable Feedstocks
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
mp
270-275 °C (dec.) (lit.)
greener alternative category
mass shift
M+5
SMILES string
[H]C(N)(Cc1c([2H])c([2H])c([2H])c([2H])c1[2H])C(O)=O
InChI
1S/C9H11NO2/c10-8(9(11)12)6-7-4-2-1-3-5-7/h1-5,8H,6,10H2,(H,11,12)/t8-/m0/s1/i1D,2D,3D,4D,5D
InChI key
COLNVLDHVKWLRT-HRVDQBBZSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service