Skip to Content
Merck
All Photos(1)

Key Documents

608793

Sigma-Aldrich

Ammonium nitrate-14N2 solution

~40 wt. % in H2O, 99.99 atom % 14N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
14NH414NO3
Molecular Weight:
80.04
UNSPSC Code:
12352116
PubChem Substance ID:

description

15N-depleted

isotopic purity

99.99 atom % 14N

concentration

~40 wt. % in H2O

bp

210 °C (lit.)

mp

169 °C (lit.)

application(s)

agriculture

mass shift

depleted

SMILES string

[14NH414NO3]

InChI

1S/HNO3.H3N/c2-1(3)4;/h(H,2,3,4);1H3

InChI key

PRORZGWHZXZQMV-UHFFFAOYSA-N

General description

Ammonium nitrate-14N2 solution is a stable isotope enriched with 14N isotope.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame over circleExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Liq. 3

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela Piccolo Maitan-Alfenas et al.
Bioresource technology, 192, 670-676 (2015-06-22)
Biomass enzymatic hydrolysis depends on the pretreatment methods employed, the composition of initial feedstock and the enzyme cocktail used to release sugars for subsequent fermentation into ethanol. In this study, sugarcane bagasse was pretreated with 1% H2SO4 and 1% NaOH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service