Skip to Content
Merck
All Photos(1)

Key Documents

606782

Sigma-Aldrich

Carbon-12C tetrafluoride

99.9 atom % 12C

Synonym(s):

Tetrafluoromethane-12C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
12CF4
Molecular Weight:
87.99
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:

description

13C-depleted

isotopic purity

99.9 atom % 12C

mass shift

depleted

SMILES string

F[12C](F)(F)F

InChI

1S/CF4/c2-1(3,4)5/i1+0

InChI key

TXEYQDLBPFQVAA-IGMARMGPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service