Skip to Content
Merck
All Photos(1)

Key Documents

604372

Sigma-Aldrich

Epichlorohydrin-2-13C

≥99 atom % 13C, ≥99% (CP), contains hydroquinone as stabilizer

Synonym(s):

2-(chloromethyl)-oxirane-13C

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC2H5ClO
CAS Number:
Molecular Weight:
93.52
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

contains

hydroquinone as stabilizer

bp

115-117 °C (lit.)

mp

-57 °C (lit.)

density

1.196 g/mL at 25 °C

mass shift

M+1

SMILES string

ClC[13CH]1CO1

InChI

1S/C3H5ClO/c4-1-3-2-5-3/h3H,1-2H2/i3+1

InChI key

BRLQWZUYTZBJKN-LBPDFUHNSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Carc. 1B - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

91.4 °F

Flash Point(C)

33 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service