Skip to Content
Merck
All Photos(1)

Key Documents

588121

Sigma-Aldrich

1-Propanol-1,1-d2

98 atom % D

Synonym(s):

Propyl-1,1-d2 alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CD2OH
CAS Number:
Molecular Weight:
62.11
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

Assay

99% (CP)

bp

97 °C (lit.)

mp

-127 °C (lit.)

density

0.830 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]C([2H])(O)CC

InChI

1S/C3H8O/c1-2-3-4/h4H,2-3H2,1H3/i3D2

InChI key

BDERNNFJNOPAEC-SMZGMGDZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

59.0 °F - closed cup

Flash Point(C)

15 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Risa Harada et al.
Journal of bioscience and bioengineering, 125(6), 688-694 (2018-01-26)
Soy sauce is a traditional Japanese fermented seasoning that contains various constituents such as amino acids, organic acids, and volatiles that are produced during the long fermentation process. Although studies regarding the correlation between microbes and aroma constituents have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service