586293
Ethanol-17O
20 atom % 17O, 99% (CP)
Synonym(s):
Ethyl alcohol-17O
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3CH217O
CAS Number:
Molecular Weight:
47.07
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
Recommended Products
isotopic purity
20 atom % 17O
Assay
99% (CP)
mol wt
46.27 by atom % calculation (20 atom % 17O)
bp
78 °C (lit.)
density
0.798 g/mL at 25 °C (lit.)
mass shift
M+1
SMILES string
CC[17OH]
InChI
1S/C2H6O/c1-2-3/h3H,2H2,1H3/i3+1
InChI key
LFQSCWFLJHTTHZ-LBPDFUHNSA-N
Packaging
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
55.4 °F - closed cup
Flash Point(C)
13 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service