Skip to Content
Merck
All Photos(1)

Key Documents

570087

Sigma-Aldrich

Melamine-d6

98 atom % D, 99% (CP)

Synonym(s):

1,3,5-Triazine-2,4,6-triamine-d6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3D6N6
CAS Number:
Molecular Weight:
132.16
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

98 atom % D

Assay

99% (CP)

mp

>300 °C (lit.)

mass shift

M+6

SMILES string

[2H]N([2H])c1nc(nc(n1)N([2H])[2H])N([2H])[2H]

InChI

1S/C3H6N6/c4-1-7-2(5)9-3(6)8-1/h(H6,4,5,6,7,8,9)/i/hD6

InChI key

JDSHMPZPIAZGSV-YIKVAAQNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

It should be noted that Melamine-d6 (570087) is an unsuitable mass spectrometric standard as it contains deuterated amine groups, which are susceptible to proton-deuterium exchange. For the usage of isotopically labeled melamine as a standard for mass spectrometry studies, please refer to Melamine-triamine-15N3 (592889).

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

572.0 °F - closed cup

Flash Point(C)

300 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service