Skip to Content
Merck
All Photos(1)

Key Documents

561142

Sigma-Aldrich

Nickel(II) bromide

anhydrous, powder, ≥99.9% trace metals basis

Synonym(s):

Nickel bromide, Nickel dibromide, Nickelous bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NiBr2
CAS Number:
Molecular Weight:
218.50
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
grade:
anhydrous
form:
powder
Pricing and availability is not currently available.

grade

anhydrous

Quality Level

Assay

≥99.9% trace metals basis

form

powder

impurities

≤1000.0 ppm Trace Metal Analysis

mp

963 °C (lit.)

density

5.098 g/mL at 25 °C (lit.)

SMILES string

Br[Ni]Br

InChI

1S/2BrH.Ni/h2*1H;/q;;+2/p-2

InChI key

IPLJNQFXJUCRNH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Nickel(II) bromide, an inorganic compound that readily dissolves in water, finds application in the manufacturing of batteries, especially as an electrode material or an electrolyte additive. It is also utilized as a catalyst or catalyst precursor in various chemical reactions, particularly in organic synthesis and industrial processes.

Application

Nickel(II) bromide can be:      
  • Used as a cathode material for sodium-metal halide batteries.
  • Used in effective passivation strategy to enhance the optical properties, efficiency, and stability of blue-emitting CsPbBr3 perovskite nanoplatelets for high-performance monochrome LED applications.
  • Used as a nickel precursor in cross-electrophile coupling reactions.   
  • Used as a precursor for the synthesis of other nickel compounds.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 1 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Questions

  1. How can the glass tube containing Nickel(II) bromide (P/N: 561142) be opened?

    1 answer
    1. Z693847 may not work on the unscored ampoules due to the tube potentially being too long. There is a specific type of opener in-house to open the ampoules

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service