Skip to Content
Merck
All Photos(1)

Key Documents

542628

Sigma-Aldrich

1,1,1-Trifluoro-2-butanone

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3COC2H5
CAS Number:
Molecular Weight:
126.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

refractive index

n20/D 1.322 (lit.)

bp

50-51 °C (lit.)

density

0.929 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

SMILES string

CCC(=O)C(F)(F)F

InChI

1S/C4H5F3O/c1-2-3(8)4(5,6)7/h2H2,1H3

InChI key

QBVHMPFSDVNFAY-UHFFFAOYSA-N

Related Categories

Application

1,1,1-Trifluoro-2-butanone may be used in the synthesis of 5α,6β-dibromo-25-hydroxycholestan-3β-yl acetate. It may also be used for the in situ generation of ethyl(trifluoromethyl)dioxirane (ETDO).[1]

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-0.0 °F - closed cup

Flash Point(C)

-17.8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoujiro Ogawa et al.
Steroids, 74(1), 81-87 (2008-11-11)
Experiments were performed to compare the regioselective hydroxylation of the isopropyl C-H bond at C-25 in 5alpha-cholestan-3beta-yl acetate by in situ generated dimethyldioxirane, methyl(trifluoromethyl)dioxirane, hexafluoro(dimethyl)dioxirane or ethyl(trifluoromethyl)dioxirane (ETDO). The dioxiranes were generated from the corresponding ketones and potassium peroxymonosulfate in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service