About This Item
Recommended Products
isotopic purity
99 atom % 13C
form
solid
optical activity
[α]20/D -40°, c = 2 in chloroform
technique(s)
mass spectrometry (MS): suitable
mp
147-149 °C (lit.)
mass shift
M+2
SMILES string
CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4[13CH2][13C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
InChI
1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1/i17+1,21+1
InChI key
HVYWMOMLDIMFJA-WKKCFSIESA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service