Skip to Content
Merck
All Photos(1)

Key Documents

474428

Sigma-Aldrich

2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl acrylate

95%

Synonym(s):

DFHA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2(CF2)5CHF2
CAS Number:
Molecular Weight:
386.13
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

contains

100 ppm 4-tert-butylcatechol as inhibitor

refractive index

n20/D 1.342 (lit.)

bp

197 °C (lit.)

density

1.581 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)COC(=O)C=C

InChI

1S/C10H6F12O2/c1-2-4(23)24-3-6(13,14)8(17,18)10(21,22)9(19,20)7(15,16)5(11)12/h2,5H,1,3H2

InChI key

QJEJDNMGOWJONG-UHFFFAOYSA-N

General description

2,2,3,3,4,4,5,5,6,6,7,7-Dodecafluoroheptyl acrylate (DFHA) is a fluorous functional monomer that is majorly used in the surface functionalization. It can also be utilized in chemical vapor deposition process for the synthesis of nanomaterials.

Application

DFHA can be used in the preparation of hybrid fluorous monolithic columns for nano-liquid chromatography. It can be used in the surface modification of multi-walled carbon nanotubes (MWCNTs) for the formation of highly conductive composites.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapor-deposited fluorinated glycidyl copolymer thin films with low surface energy and improved mechanical properties
Mao Y and Gleason KK
Macromolecules, 39(11), 3895-3900 (2006)
One-pot two-step perfluoroalkylsilane functionalization of multi-walled carbon nanotubes for polyurethane-based composites
Li S, et al.
Composites Science and Technology, 143, 46-55 (2017)
Haiyang Zhang et al.
Journal of chromatography. A, 1440, 66-73 (2016-02-27)
A hybrid fluorous monolithic column was simply prepared via photo-initiated free radical polymerization of an acrylopropyl polyhedral oligomeric silsesquioxane (acryl-POSS) and a perfluorous monomer (2,2,3,3,4,4,5,5,6,6,7,7-dodecafluoroheptyl acrylate) in UV-transparent fused-silica capillaries within 5min. The physical characterization of hybrid fluorous monolith, including

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service