Skip to Content
Merck
All Photos(2)

Documents

469963

Sigma-Aldrich

L-Threoninol

97%, for peptide synthesis

Synonym(s):

(2R,3R)-2-Amino-1,3-butanediol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)CH(NH2)CH2OH
CAS Number:
Molecular Weight:
105.14
Beilstein:
6130524
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

product name

L-Threoninol, 97%

Quality Level

Assay

97%

form

solid

optical activity

[α]20/D −4.2°, c = 1% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

49-54 °C (lit.)

application(s)

peptide synthesis

SMILES string

C[C@@H](O)[C@H](N)CO

InChI

1S/C4H11NO2/c1-3(7)4(5)2-6/h3-4,6-7H,2,5H2,1H3/t3-,4-/m1/s1

InChI key

MUVQIIBPDFTEKM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

L-Threoninol also known as (2R,3R)-2-Amino-1,3-butanediol, is an amino alcohol, which is commonly used in solution phase peptide synthesis.

Application

L-Threoninol can be used as artificial abasic nucleoside in the synthesis and modification of oligodeoxynucleotide and can also be utilized to synthesize pyrene-L-threoninyl analogues.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potent triple helix stabilization by 5?, 3?-modified triplex-forming oligonucleotides
N Ben Gaied
Chembiochem, 10, 1839-1851 (2009)
Synthesis and properties of an oligonucleotide modified with an acridine derivative at the artificial abasic site
K Fukui
Bioconjugate Chemistry, 7, 349-355 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service