Skip to Content
Merck
All Photos(2)

Key Documents

463442

Sigma-Aldrich

5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide

95%

Synonym(s):

4-[2-(5-Chloro-2-methoxybenzamido)ethyl]phenylsulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(OCH3)CONHCH2C6H4SO2NH2
CAS Number:
Molecular Weight:
368.84
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

95%

mp

209-214 °C (lit.)

functional group

amide
chloro

SMILES string

COc1ccc(Cl)cc1C(=O)NCCc2ccc(cc2)S(N)(=O)=O

InChI

1S/C16H17ClN2O4S/c1-23-15-7-4-12(17)10-14(15)16(20)19-9-8-11-2-5-13(6-3-11)24(18,21)22/h2-7,10H,8-9H2,1H3,(H,19,20)(H2,18,21,22)

InChI key

KVWWTCSJLGHLRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide is an organic building block. It has been reported as an intermediate in the synthesis of glyburide.[1] Synthesis of 5-chloro-2-methoxy-N-[2-(4-sulfamoylphenyl)ethyl]benzamide has been reported.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular regulation of cell fate in cerebral ischemia: role of the inflammasome and connected pathways
Trendelenburg G.
Journal of Cerebral Blood Flow and Metabolism (2014)
Reply to Letter Regarding Article," NLRP3 Inflammasome as a Therapeutic Target in Myocardial Infarction.
Takahashi M.
International Heart Journal, 55(4), 380-380 (2014)
Carlo Marchetti et al.
Journal of cardiovascular pharmacology, 63(4), 316-322 (2013-12-18)
The formation of the NLRP3 inflammasome in the heart during acute myocardial infarction amplifies the inflammatory response and mediates further damage. Glyburide has NLRP3 inhibitory activity in vitro but requires very high doses in vivo, associated with hypoglycemia. The aim

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service