Skip to Content
Merck
All Photos(1)

Documents

454265

Sigma-Aldrich

2′,5′-Dimethylacetophenone

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C6H3COCH3
CAS Number:
Molecular Weight:
148.20
Beilstein:
2042139
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.529 (lit.)

bp

195 °C (lit.)

density

0.988 g/mL at 25 °C (lit.)

SMILES string

CC(=O)c1cc(C)ccc1C

InChI

1S/C10H12O/c1-7-4-5-8(2)10(6-7)9(3)11/h4-6H,1-3H3

InChI key

AWKBVLVKQQRRFQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Orientation in the Nitration of 3, 4-and 2, 5-Dimethylacetophenone.
Buu-Hoi NP, et al.
The Journal of Organic Chemistry, 17(7), 1000-1004 (1952)
Assessment of substituent effects and antimicrobial activities of some 2', 5'-dimethyl phenyl chalcones.
Joseph SJ, et al.
International Letters of Chemistry, Physics and Astronomy, 5, 99-123 (2014)
Photoenolization of ortho-alkyl-substituted acetophenones: evidence for the enol triplet state.
Findlay DM and Tchir MF.
J. Chem. Soc., Faraday, 1096-1100 (1976)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service