Skip to Content
Merck
All Photos(2)

Documents

452718

Sigma-Aldrich

Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine

Dye content 90 %

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H24N8Zn
CAS Number:
Molecular Weight:
682.06
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

88.5% (HPLC)

Quality Level

form

powder

composition

Dye content, 90%

mp

>300 °C (lit.)

λmax

423 nm

ε (extinction coefficient)

≥200000 at 420-426 nm in chloroform

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Zn]1n2c3ccc2c(-c4ccncc4)c5ccc(n5)c(-c6ccncc6)c7ccc(c(-c8ccncc8)c9ccc(n9)c3-c%10ccncc%10)n17

InChI

1S/C40H24N8.Zn/c1-2-30-38(26-11-19-42-20-12-26)32-5-6-34(47-32)40(28-15-23-44-24-16-28)36-8-7-35(48-36)39(27-13-21-43-22-14-27)33-4-3-31(46-33)37(29(1)45-30)25-9-17-41-18-10-25;/h1-24H;/q-2;+2/b37-29-,37-31-,38-30-,38-32-,39-33-,39-35-,40-34-,40-36-;

InChI key

PZGANULLXCGPFZ-GFPSKMGBSA-N

Application

Zinc 5,10,15,20-tetra(4-pyridyl)-21H,23H-porphine (ZnTPyP) forms an organic thin-film, which can be used to fabricate hybrid heterojunction solar cells. It is also used in the production of organic nanostructures, which find applications in organic field-effect transistors (OFET), organic light-emitting displays (OLED), and nanosensors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service