Skip to Content
Merck
All Photos(1)

Key Documents

445886

Sigma-Aldrich

Poly(ethylene glycol) dimethyl ether

average MN 500, cross-linking reagent click chemistry, 100 ppm BHT as stabilizer

Synonym(s):

Polyethylene glycol, Dimethyl polyethylene glycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O(CH2CH2O)nCH3
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Product Name

Poly(ethylene glycol) dimethyl ether, average Mn ~500, contains 100 ppm BHT as stabilizer

form

liquid

Quality Level

mol wt

average Mn ~500

contains

100 ppm BHT as stabilizer

reaction suitability

reagent type: cross-linking reagent
reaction type: click chemistry

bp

>250 °C

density

1.05 g/mL at 25 °C

polymer architecture

shape: linear
functionality: homobifunctional

SMILES string

CO.OCCO

InChI

1S/C4H10O2/c1-5-3-4-6-2/h3-4H2,1-2H3

InChI key

XTHFKEDIFFGKHM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Mechanically and Thermally Robust Gel Electrolytes Built from A Charged Double Helical Polymer.: This research describes the development of gel electrolytes incorporating Poly(ethylene glycol) dimethyl ether (PEGDME), highlighting their enhanced mechanical and thermal stability, which are crucial for high-performance energy storage applications (Yu et al., 2024).
  • An environmentally benign protocol for the synthesis of sugar 1,2-orthoesters in poly(ethylene glycol) dimethyl ether (DMPE).: This study presents a green synthesis approach using PEGDME as a reaction medium for producing sugar orthoesters, demonstrating its role in sustainable chemical processes within the pharmaceutical industry (Yang et al., 2023).
  • Room-Temperature Solid-State Polymer Electrolyte in Li-LiFePO(4) , Li-S and Li-O(2) Batteries.: This article details the use of PEGDME-based polymer electrolytes in various lithium battery configurations, emphasizing its effectiveness in enhancing battery safety and performance (Marangon et al., 2023).
  • Safety, high-performing and effects of the N/P ratio of a solid lithium ion battery using PEGDME based polymer electrolytes.: Discusses the implementation of PEGDME in solid lithium-ion batteries, focusing on its role in achieving high performance and safety, critical for the advancement of energy storage technologies (Jeon et al., 2023).

Analysis Note

Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

312.8 °F - closed cup

Flash Point(C)

156.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yimu Zhao et al.
Advanced healthcare materials, 8(5), e1801187-e1801187 (2019-02-10)
Due to escalating drug developmental costs and limitations of cardiotoxicity screening, there is an urgent need to develop robust in vitro 3D tissue culture platforms that can both facilitate the culture of human cardiac tissues and provide noninvasive functional readouts

Questions

  1. What is the viscosity of this material?

    1 answer
    1. 445886 is Poly(ethylene glycol) dimethyl ether. Unfortunately, the viscosity is not tested as per its COA: https://www.sigmaaldrich.com/certificates/sapfs/PROD/sap/certificate_pdfs/COFA/Q14/445886-BULKBCCL5083.pdf.
      The viscosity has been reported by some research papers. For example, Here is the research paper that reported the dynamic viscosity is 5.8 mPa·s at 298 K.
      Li, Yifu; Chemical Engineering and Processing, (2017), 121, 149-161.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service