Skip to Content
Merck
All Photos(2)

Key Documents

43749

Sigma-Aldrich

Ethyl 1,3-dithiane-2-carboxylate

technical, ≥90% (GC)

Synonym(s):

Glyoxylic acid ethyl ester trimethylenemercaptal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12O2S2
CAS Number:
Molecular Weight:
192.30
Beilstein:
1424352
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (GC)

refractive index

n20/D 1.539 (lit.)
n20/D 1.541

bp

75-77 °C/0.2 mmHg (lit.)

density

1.22 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCOC(=O)C1SCCCS1

InChI

1S/C7H12O2S2/c1-2-9-6(8)7-10-4-3-5-11-7/h7H,2-5H2,1H3

InChI key

ANEDZEVDORCLPM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethyl 1,3-dithiane-2-carboxylate is an α-keto acid equivalent and bulky equivalent of acetate. It participates in syn-selective aldol reactions. It can be prepared from the reaction of ethyl diethoxyacetate and 1,3-propanedithiol in the presence of BF3/Et2O. Asymmetric oxidation of ethyl 1,3-dithiane-2-carboxylate by Modena protocol has been reported to afford trans bis-sulfoxide in 60% yield. Carbanion from ethyl 1,3-dithiane-2-carboxylate may be employed for the preparation of α-keto esters.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alkylation of the carbanion from methyl bis (ethylthio) acetate with alkyl and aralkyl halides.
Lerner LM.
The Journal of Organic Chemistry, 41(12), 2228-2229 (1976)
Convenient synthesis of α-keto esters.
Eliel EL and Hartmann AA.
The Journal of Organic Chemistry, 37(3), 505-506 (1972)
Varinder K. Aggarwal et al.
The Journal of organic chemistry, 63(21), 7306-7310 (2001-10-24)
Asymmetric oxidation of a range of 1,3-dithianes was studied using the Kagan protocol [CHP (4 equiv), (+)-DET (2 equiv), Ti(OiPr)(4) (1 equiv), and H(2)O (1 equiv) at -35 degrees C for 48 h]. 1,3-Dithiane itself gave monoxide (30% ee) and
Dianions of glyoxylic acid thioketals: conventent α-keto acid equivalents.
Bates GS and Ramaswamy S.
Canadian Journal of Chemistry, 58(7), 716-722 (1980)
L.A. Flippin et al.
Tetrahedron Letters, 26, 2977-2977 (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service