Skip to Content
Merck
All Photos(1)

Key Documents

425540

Sigma-Aldrich

Boron subphthalocyanine chloride

Dye content ~85 %

Synonym(s):

Boron(III) subphthalocyanine chloride, Chloro(subphthalocyaninato)boron, Chloroboronsubphthalocyanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H12BClN6
CAS Number:
Molecular Weight:
430.66
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

composition

Dye content, ~85%

mp

>360 °C (lit.)

λmax

565 nm

OPV Device Performance

ITO/MoO3/SubPc/F16CuPc/BCP/Al

  • Short-circuit current density (Jsc): 2.54 mA/cm2
  • Open-circuit voltage (Voc): 0.4 V
  • Fill Factor (FF): 0.55
  • Power Conversion Efficiency (PCE): 0.56 %

ITO/SubPc/F16CuPc/Al
  • Short-circuit current density (Jsc): 0.46 mA/cm2
  • Open-circuit voltage (Voc): 0.22 V
  • Fill Factor (FF): 0.21
  • Power Conversion Efficiency (PCE): 0.02 %

ITO/SubPc/F16CuPc/BCP/Al
  • Short-circuit current density (Jsc): 1.9 mA/cm2
  • Open-circuit voltage (Voc): 0.21 V
  • Fill Factor (FF): 0.31
  • Power Conversion Efficiency (PCE): 0.13 %

SMILES string

Clb1n2c3nc4nc(nc5c6ccccc6c(nc2c7ccccc37)n15)c8ccccc48

InChI

1S/C24H12BClN6/c26-25-31-21-15-9-3-5-11-17(15)23(31)30-24-18-12-6-4-10-16(18)22(32(24)25)29-20-14-8-2-1-7-13(14)19(27-20)28-21/h1-12H/b28-19-,28-21-,29-20-,29-22-

InChI key

IFJKCEPOUBWFFD-QJAIGJGMSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service