Skip to Content
Merck
All Photos(1)

Documents

405663

Sigma-Aldrich

Diphenyl(2,4,6-trimethylbenzoyl)phosphine oxide/2-hydroxy-2-methylpropiophenone, blend

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.475 (lit.)

density

1.12 g/mL at 25 °C (lit.)

SMILES string

CC(C)(O)C(=O)c1ccccc1.Cc2cc(C)c(c(C)c2)C(=O)P(=O)(c3ccccc3)c4ccccc4

InChI

1S/C22H21O2P.C10H12O2/c1-16-14-17(2)21(18(3)15-16)22(23)25(24,19-10-6-4-7-11-19)20-12-8-5-9-13-20;1-10(2,12)9(11)8-6-4-3-5-7-8/h4-15H,1-3H3;3-7,12H,1-2H3

InChI key

SEHZJEJTNRNDFD-UHFFFAOYSA-N

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service