Skip to Content
Merck
All Photos(3)

Key Documents

403229

Sigma-Aldrich

4′-Bromo-2,2,2-trifluoroacetophenone

≥98%

Synonym(s):

4-Bromo-α,α,α-trifluoroacetophenone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4COCF3
CAS Number:
Molecular Weight:
253.02
Beilstein:
1956460
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

bp

95 °C/4 mmHg (lit.)

mp

26-30 °C (lit.)

density

1.662 g/mL at 25 °C (lit.)

functional group

bromo
fluoro
ketone

SMILES string

FC(F)(F)C(=O)c1ccc(Br)cc1

InChI

1S/C8H4BrF3O/c9-6-3-1-5(2-4-6)7(13)8(10,11)12/h1-4H

InChI key

IHGSAQHSAGRWNI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

4′-Bromo-2,2,2-trifluoroacetophenone may be used in the preparation of carbonyl-bridged bithiazole derivatives.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Air-Stable n-Type Organic Field-Effect Transistors Based on Carbonyl-Bridged Bithiazole Derivatives.
Ie Y, et al.
Advances in Functional Materials, 20(6), 907-913 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service