Skip to Content
Merck
All Photos(2)

Documents

398411

Sigma-Aldrich

Gold(I) iodide

99.9% trace metals basis

Synonym(s):

Gold monoiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AuI
CAS Number:
Molecular Weight:
323.87
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: gold

impurities

≤1500.0 ppm Trace Metal Analysis

mp

120 °C (dec.) (lit.)

density

8.25 g/mL at 25 °C (lit.)

SMILES string

I[Au]

InChI

1S/Au.HI/h;1H/q+1;/p-1

InChI key

ATGIETUGWDAYPU-UHFFFAOYSA-M

General description

AuI can be prepared by adding KI in an acidic solution of HAuCl4.

Application

Gold(I) iodide may be used as a catalyst in Sonogashira reactions, wherein terminal alkynes were reacted with aryl iodides and bromides to yield corresponding cross coupling products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold (I) iodide catalyzed Sonogashira reactions.
Li P, et al.
European Journal of Organic Chemistry, 35, 5946-5951 (2008)
Synth. Meth. Organomet. Inorg. Chem., 60-60 (1997)

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service