Skip to Content
Merck
All Photos(1)

Key Documents

365815

Sigma-Aldrich

4-(Trifluoromethyl)benzyl chloride

98%

Synonym(s):

α′-Chloro-α,α,α-trifluoro-p-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4CH2Cl
CAS Number:
Molecular Weight:
194.58
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.464 (lit.)

bp

68 °C/12 mmHg (lit.)

density

1.315 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1ccc(CCl)cc1

InChI

1S/C8H6ClF3/c9-5-6-1-3-7(4-2-6)8(10,11)12/h1-4H,5H2

InChI key

MCHDHQVROPEJJT-UHFFFAOYSA-N

General description

Electrochemical reduction of 4-(trifluoromethyl)benzyl chloride catalyzed by Co(salen) (H2salen, N,N′-bis(salicylidene)-ethane-1,2-diamine) in acetonitrile is reported.

Application

4-(Trifluoromethyl)benzyl chloride may be used in the synthesis of novel series of dithiocarbamates, via reaction with sodium salts of N,N-disubstituted dithiocarbamic acids.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanism of the electrochemical reduction of benzyl chlorides catalysed by Co (salen).
Isse AA, et al.
Journal of Electroanalytical Chemistry, 444(2), 241-245 (1992)
Mehlika D Altıntop et al.
Archiv der Pharmazie, 346(8), 571-576 (2013-07-25)
In the present paper, a novel series of dithiocarbamates was synthesized via the treatment of 4-(trifluoromethyl)benzyl chloride with appropriate sodium salts of N,N-disubstituted dithiocarbamic acids. The chemical structures of the compounds were elucidated by (1) H NMR, mass spectral data
Nicholas R Oranzi et al.
Analytical chemistry, 91(6), 4092-4099 (2019-02-27)
Quantitation of the serum concentration of 25-hydroxyvitamin D is a high-demand assay that suffers from long chromatography time to separate 25-hydroxyvitamin D from its inactive epimer; however, ion mobility spectrometry can distinguish the epimer pair in under 30 ms due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service