Skip to Content
Merck
All Photos(1)

Key Documents

278971

Sigma-Aldrich

Diphenyl N-cyanocarbonimidate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCN=C(OC6H5)2
CAS Number:
Molecular Weight:
238.24
Beilstein:
4254248
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

155-158 °C (lit.)

functional group

amine
phenoxy

SMILES string

N#C\N=C(/Oc1ccccc1)Oc2ccccc2

InChI

1S/C14H10N2O2/c15-11-16-14(17-12-7-3-1-4-8-12)18-13-9-5-2-6-10-13/h1-10H

InChI key

SLIKWVTWIGHFJE-UHFFFAOYSA-N

Gene Information

human ... EPHX2(2053)
mouse ... Ephx2(13850)

Looking for similar products? Visit Product Comparison Guide

Application

Diphenyl N-cyanocarbonimidate has been used in the preparation of:
  • N-substituted 2-amino-4H-3,1-benzoxazines
  • cyanoguanidine derivatives
  • oxadiazoles

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

4H-3, 1-Benzoxazines, Their Salts and Dihydro Derivatives.(Review).
Gromachevskaya EV, et al.
Chemistry of Heterocyclic Compounds, 39(2), 137-155 (2003)
Bassem Sadek et al.
Molecules (Basel, Switzerland), 18(11), 14186-14202 (2013-11-20)
Premedication with a combination of histamine H₁ receptor (H₁R) and H₂ receptor (H₂R) antagonists has been suggested as a prophylactic principle, for instance, in anaesthesia and surgery. Aiming at pharmacological hybrids combining H₁R and H₂R antagonistic activity, a series of
Tetrahedron Letters, 48, 2733-2733 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service