Skip to Content
Merck
All Photos(1)

Key Documents

252492

Sigma-Aldrich

Benzyl isothiocyanate

98%

Synonym(s):

Benzyl mustard oil, Isothiocyanotaomethylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2NCS
CAS Number:
Molecular Weight:
149.21
Beilstein:
386135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.601 (lit.)

bp

242-243 °C (lit.)

density

1.125 g/mL at 25 °C (lit.)

functional group

amine
isothiocyanate
phenyl

storage temp.

2-8°C

SMILES string

S=C=NCc1ccccc1

InChI

1S/C8H7NS/c10-7-9-6-8-4-2-1-3-5-8/h1-5H,6H2

InChI key

MDKCFLQDBWCQCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzyl isothiocyanate is a naturally-occurring constituent of cruciferous vegetables[1]. It has antibacterial properties and its metabolism in man has been investigated[2]. It inhibits chemically induced cancer in animal models[1].

Application

Benzyl isothiocyanate can be used as a reactant to synthesize:
  • N

  • -benzylthioureas by reacting with various amines.[3]
  • N-benzyl-O-alkyl carbamates by treating with long-chain alcohols.[4]
  • 3-mercapto-1,2,4-triazole building block by reacting with formylhydrazine via acyl thiosemicarbazide intermediate formation.[5]
  • S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine.[6]

Benzyl isothiocyanate has been used in the preparation of S-(N-benzylthiocarbamoyl)-L-glutathione and S-(N-benzylthiocarbamoyl)-L-cysteine[6].

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

222.8 °F

Flash Point(C)

106 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

G Brüsewitz et al.
The Biochemical journal, 162(1), 99-107 (1977-01-15)
1. The corresponding cysteine conjugate was formed when the GSH (reduced glutathione) or cysteinylglycine conjugates of benzyl isothiocyanate were incubated with rat liver or kidney homogenates. When the cysteine conjugate of benzyl isothiocyanate was similarly incubated in the presence of
Synthesis of 5-substituted 3-mercapto-1, 2, 4-triazoles via Suzuki-Miyaura reaction
Katkevica S, et al.
Tetrahedron Letters, 54(34), 4524-4525 (2013)
A Simple and Green Procedure for the Synthesis of N-Benzylthioureas
C de Sequeira Aguiar L, et al.
Letters in Organic Chemistry, 8(8), 540-544 (2011)
Su-Hua Huang et al.
Journal of agricultural and food chemistry, 60(2), 665-675 (2011-12-14)
Benzyl isothiocyanates (BITC), a member of the isothiocyanate (ITC) family, inhibits cell growth and induces apoptosis in many types of human cancer cell lines. The present study investigated mechanisms underlying BITC-induced apoptosis in A375.S2 human melanoma cancer cells. To observe
Srinivas Reddy Boreddy et al.
PloS one, 6(10), e25799-e25799 (2011-10-22)
Our previous studies have shown that benzyl isothiocyanate (BITC) suppresses pancreatic tumor growth by inhibiting STAT-3; however, the exact mechanism of tumor growth suppression was not clear. Here we evaluated the effects and mechanism of BITC on pancreatic tumor angiogenesis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service