Skip to Content
Merck
All Photos(1)

Key Documents

209880

Sigma-Aldrich

5-Phenyl-o-anisidine

≥98%

Synonym(s):

2-Methoxy-5-phenylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H3(OCH3)NH2
CAS Number:
Molecular Weight:
199.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

mp

82-83 °C (lit.)

functional group

phenyl

SMILES string

COc1ccc(cc1N)-c2ccccc2

InChI

1S/C13H13NO/c1-15-13-8-7-11(9-12(13)14)10-5-3-2-4-6-10/h2-9H,14H2,1H3

InChI key

DTYBRSLINXBXMP-UHFFFAOYSA-N

General description

5-Phenyl-o-anisidine is a promising monomer for polyanilines studies. X-ray Diffraction and DFT studies of 5-phenyl-o-anisidine (2-methoxy-5-phenylaniline) have been reported.

Application

5-Phenyl-o-anisidine (2-Methoxy-5-phenylaniline) was used in the synthesis of 2,4-diamino-5-methyl-6-substituted-pyrido[2,3-d]pyrimidines.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Gangjee et al.
Journal of medicinal chemistry, 40(4), 479-485 (1997-02-14)
Nine novel 2,4-diamino-5-methyl-6-substituted-pyrido[2,3-d]pyrimidines, 2-10, were synthesized as potential inhibitors of Pneumocystis carinii dihydrofolate reductase (pcDHFR) and Toxoplasma gondii dihydrofolate reductase (tgDHFR). Compounds 2-5 were designed as conformationally restricted analogues of trimetrexate (TMQ), in which rotation around tau 3 was constrained
X-ray Diffraction and DFT Studies of 2-Methoxy-5-phenylaniline.
Marques AT, et al.
Journal of Chemical Crystallography, 38(4), 295-299 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service