Skip to Content
Merck
All Photos(1)

Key Documents

196975

Sigma-Aldrich

5-(Trifluoromethyl)-1,3-phenylenediamine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H3(NH2)2
CAS Number:
Molecular Weight:
176.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

87-89 °C (lit.)

functional group

fluoro

SMILES string

Nc1cc(N)cc(c1)C(F)(F)F

InChI

1S/C7H7F3N2/c8-7(9,10)4-1-5(11)3-6(12)2-4/h1-3H,11-12H2

InChI key

KZSXRDLXTFEHJM-UHFFFAOYSA-N

Related Categories

Application

5-(Trifluoromethyl)-1,3-phenylenediamine was used in the synthesis of:
  • hybrid materials based on new polyhedral oligomeric silsesquioxane
  • fluorinated aromatic polyimide films

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Organic-inorganic hybrid materials from a new octa (2, 3-epoxypropyl) silsesquioxane with diamines.
Lee L-H and Chen W-C.
Polymer, 46(7), 2163-2174 (2005)
Preparation of microporous carbon films from fluorinated aromatic polyimides.
Ohta N, et al.
Carbon, 46(10), 1350-1357 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service