Skip to Content
Merck
All Photos(1)

Key Documents

158992

Sigma-Aldrich

Methyl p-toluenesulfonate

98%

Synonym(s):

Methyl p-methylbenzenesulfonate, Methyl toluene-4-sulfonate, Methyl tosylate, Methylp-tosylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO3CH3
CAS Number:
Molecular Weight:
186.23
Beilstein:
609209
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

6.45 (vs air)

Quality Level

vapor pressure

1 mmHg ( 20 °C)

Assay

98%

form

solid

autoignition temp.

896 °F

refractive index

n20/D 1.5172 (lit.)

bp

144-145 °C/5 mmHg (lit.)

mp

25-28 °C (lit.)

density

1.234 g/mL at 25 °C (lit.)

functional group

tosylate

storage temp.

2-8°C

SMILES string

COS(=O)(=O)c1ccc(C)cc1

InChI

1S/C8H10O3S/c1-7-3-5-8(6-4-7)12(9,10)11-2/h3-6H,1-2H3

InChI key

VUQUOGPMUUJORT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl p-toluenesulfonate is sulfonate ester present as potentially genotoxic impurity in drug substances. Kinetics of reactions of methyl p-toluenesulfonate with N,N-dimethylaniline has been investigated. It participates in selective 1-substitution reaction of tetrazole.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Secondary deuterium isotope effects.. beta.-Kinetic effects in Sn2 reactions of N, N-dimethylaniline and dimethylphenylphosphine with methyl p-toluenesulfonate, and comparison with observed and calculated vibrational frequencies.
Kaplan ED and Thornton ER.
Journal of the American Chemical Society, 89(25), 6644-6651 (1967)
The formation of tin-nitrogen bonds. V. The selective 1-substitution reaction of tetrazoles by the reaction of 5-substituted 2-(tri-n-butylstannyl) tetrazoles with methyl iodide, methyl p-toluenesulfonate, dimethyl sulfate, and ethyl bromoacetate.
Isida T, et al.
Bulletin of the Chemical Society of Japan, 46, 2176-2180 (1973)
Joachim F R Van Guyse et al.
Polymers, 13(3) (2021-02-04)
Smart or adaptive materials often utilize stimuli-responsive polymers, which undergo a phase transition in response to a given stimulus. So far, various stimuli have been used to enable the modulation of drug release profiles, cell-interactive behavior, and optical and mechanical
Bryn D Monnery et al.
International journal of pharmaceutics, 521(1-2), 249-258 (2017-02-25)
The mechanism of polycation cytotoxicity and the relationship to polymer molecular weight is poorly understood. To gain an insight into this important phenomenon a range of newly synthesised uniform (near monodisperse) linear polyethylenimines, commercially available poly(l-lysine)s and two commonly used
Polen Kocak et al.
Macromolecular bioscience, 21(2), e2000287-e2000287 (2020-11-17)
Prostate cancer is the most common cancer, which is about 15-20% among male cancers worldwide. As most common strategies such as radiotherapy, chemotherapy, or surgery alone can be unsuccessful in the treatment of prostate cancer, this study aims to develop

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service