Skip to Content
Merck
All Photos(2)

Key Documents

143456

Sigma-Aldrich

2-Amino-5-chlorothiazole hydrochloride

95%

Synonym(s):

(5-Chlorothiazol-2-yl)amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H3ClN2S · HCl
CAS Number:
Molecular Weight:
171.05
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

164-167 °C (dec.) (lit.)

SMILES string

Cl[H].Nc1ncc(Cl)s1

InChI

1S/C3H3ClN2S.ClH/c4-2-1-6-3(5)7-2;/h1H,(H2,5,6);1H

InChI key

GTMGFQYVLSQTKP-UHFFFAOYSA-N

Application

2-Amino-5-chlorothiazole hydrochloride was used as starting reagent in the synthesis of 2-chloro-6-methylimidazo[2,1-b]thiazole.
  • Reactant for preparation of biologically active thiazole derivatives

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aldo Andreani et al.
Bioorganic & medicinal chemistry, 12(21), 5525-5532 (2004-10-07)
In this work we describe the synthesis of a series of imidazo[2,1-b]thiazoles and 2,3-dihydroimidazo[2,1-b]thiazoles connected by means of a methylene bridge to CoQ(0). These compounds were tested as specific inhibitors of the NADH:ubiquinone reductase activity in mitochondrial membranes. The imidazothiazole

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service