Skip to Content
Merck
All Photos(1)

Documents

131946

Sigma-Aldrich

2,2,4,4-Tetramethyl-1,3-cyclobutanediol, mixture of isomers

99%

Synonym(s):

1,3-Dihydroxy-2,2,4,4-tetramethylcyclobutane, 2,2,4,4-Tetramethyl-1,3-cyclobutandiol, 2,2,4,4-Tetramethyl-1,3-cyclobutanediol, 2,4-Dihydroxy-1,1,3,3-tetramethylcyclobutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4C4H2(OH)2
CAS Number:
Molecular Weight:
144.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

210-215 °C (lit.)

mp

126-129 °C (lit.)

SMILES string

CC1(C)C(O)C(C)(C)C1O

InChI

1S/C8H16O2/c1-7(2)5(9)8(3,4)6(7)10/h5-6,9-10H,1-4H3

InChI key

FQXGHZNSUOHCLO-UHFFFAOYSA-N

Application

2,2,4,4-Tetramethyl-1,3-cyclobutanediol was used in the preparation of Eastman Tritan copolyester.

Legal Information

Tritan is a trademark of Eastman Kodak Company

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Sol. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ju-Won Jang et al.
Environmental pollution (Barking, Essex : 1987), 259, 113907-113907 (2020-02-07)
B cells contribute to produce inflammatory cytokines and antibodies, to present autoantigens, and to interact with T cells, which lead to body defense and disease control. Nuclear factor (erythroid-derived 2)-like 2(Nrf2) is responsible for gene expression of antioxidant enzymes to
Thomas G Osimitz et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 50(6), 2196-2205 (2012-02-22)
Eastman Tritan™ copolyester, a novel plastic from Eastman is manufactured utilizing three monomers, di-methylterephthalate (DMT), 1,4-cyclohexanedimethanol (CHDM), and 2,2,4,4-tetramethyl-1,3-cyclobutanediol (TMCD) in various ratios. As with most any polymer, the monomers along with the high molecular weight oligomers, whose toxicity is
Fabrian Brenz et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 34(2), 307-319 (2016-11-29)
Polyesters (PESs) are gaining more importance on the food contact material (FCM) market and the variety of properties and applications is expected to be wide. In order to acquire the desired properties manufacturers can combine several FCM-approved polyvalent carboxylic acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service