Skip to Content
Merck
All Photos(1)

Key Documents

117277

Sigma-Aldrich

Bis(4-bromophenyl) ether

99%

Synonym(s):

4-Bromophenyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(BrC6H4)2O
CAS Number:
Molecular Weight:
328.00
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

99%

form

solid

bp

338-340 °C (lit.)

mp

61-63 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc(Oc2ccc(Br)cc2)cc1

InChI

1S/C12H8Br2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H

InChI key

YAWIAFUBXXPJMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis-(4-bromophenyl)-ether has been used in the preparation of 4-[4′-(diethoxyphosphoryl)phenoxy]phenyl-phosphonic acid diethyl ester[1].

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Mar Gómez-Alcántara et al.
Inorganic chemistry, 43(17), 5283-5293 (2004-08-18)
Commercial bis-(4-bromophenyl)-ether, [BrC(6)H(4)](2)-O, has been used to prepare 4-[4'-(diethoxyphosphoryl)phenoxy]phenyl-phosphonic acid diethyl ester, [(CH(3)CH(2))(2)O(3)P-C(6)H(4)](2)-O, (I) following a slight modification of the Michaelis-Arbuzov reaction. The acid hydrolysis of I gave 4-(4'-phosphonophenoxy)phenyl phosphonic acid, [H(2)O(3)P-C(6)H(4)](2)-O (II), and both compounds have been characterized by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service