Skip to Content
Merck
All Photos(1)

Key Documents

PHL86543

Arnicolide C

phyproof® Reference

Synonym(s):

11,13-Dihydrohelenalin isobutyrate, 6-O-Isobutyryl 11,13-dihydrohelenalin, Isobutyroylplenolin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H26O5
CAS Number:
Molecular Weight:
334.41
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages
pharmaceutical

storage temp.

2-8°C

SMILES string

O1[C@H]2[C@H]([C@@H]([C@]3([C@H]([C@@H](C2)C)C=CC3=O)C)OC(=O)C(C)C)[C@@H](C1=O)C

Looking for similar products? Visit Product Comparison Guide

General description

This substance is supplied with certified chromatographic purity. The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Other Notes

This compound is commonly found in plants of the genus: arnica

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service