Skip to Content
Merck
All Photos(1)

Key Documents

89013

Supelco

Myricetin

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

3,3′,4′,5,5′,7-Hexahydroxyflavone, Cannabiscetin, Myricetol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O8
CAS Number:
Molecular Weight:
318.24
Beilstein:
332331
MDL number:
UNSPSC Code:
12352202

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

expiry date on the label
limited shelf life

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

mp

>300 °C (lit.)

application(s)

food and beverages

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C(=O)C(O)=C(Oc2c1)c3cc(O)c(O)c(O)c3

InChI

1S/C15H10O8/c16-6-3-7(17)11-10(4-6)23-15(14(22)13(11)21)5-1-8(18)12(20)9(19)2-5/h1-4,16-20,22H

InChI key

IKMDFBPHZNJCSN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Other Notes

This compound is commonly found in plants of the genus: eucalyptus hypericum primula

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service