Skip to Content
Merck
All Photos(1)

Key Documents

814691

TOYOPEARL® HW-75F Bulk Media

phase hydroxylated methacrylate, bottle of 1 L, 45 μm particle size

Synonym(s):

TOYOPEARL® HW-75 Size Exclusion Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
particle size:
45 μm
matrix active group:
hydroxylated methacrylate phase
matrix:
hydroxylated methacrylic polymer

product line

TOYOPEARL®

form

slurry

packaging

bottle of 1 L

manufacturer/tradename

Tosoh 14691

parameter

3 bar max. pressure

technique(s)

LPLC: suitable

matrix

hydroxylated methacrylic polymer

matrix active group

hydroxylated methacrylate phase

particle size

45 μm

operating pH

2-13

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl HW-75 resin is a hydroxylated methacrylic polymer bead for size exclusion chromatography of proteins between 500,000-50,000,000 Da.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

72% (v/v) slurry in 20% (v/v) ethanol
Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service