Skip to Content
Merck
All Photos(1)

Key Documents

59629

Supelco

SUPELCOSIL LC-18-S (5 µm) HPLC Columns

L × I.D. 2 cm × 4 mm Supelguard Guard Cartridge pkg of 2 ea, Guard Cartridge holder required for use

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700

Product Name

SUPELCOSIL LC-18-S Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm

Agency

suitable for USP L1

description

Supelguard Cartridge

packaging

pkg of 1 kit

technique(s)

HPLC: suitable

L × I.D.

2 cm × 4 mm

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

120 Å

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

SUPELCOSIL LC-18-S Supelguard Cartridges [5 μm particle size, L × I.D. 2 cm × 4 mm] contain a SUPELCOSIL packing, in a 2 cm stainless steel body, enclosed by polyether ether ketone (PEEK) encapsulated stainless steel frits (2 μm porosity). A Supelguard kit (one cartridge, a stand-alone holder, tubing, and 2 nuts and ferrules) enables one to use the cartridge with any analytical column. A direct-connect guard cartridge holder can directly connect a Supelguard cartridge to a Supelco analytical column. 3.0 mm cartridges are available in packs of two only-purchase a stand-alone or modular holder separately. Low volume 3.0 mm ID cartridges are a good choice for protecting an analytical column containing 3 μm particles.

Footnote

Kit includes one cartridge, a stand-alone holder, a piece of tubing, and 2 nuts and ferrules.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

SUPELCOSIL is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Crozatier et al.
Molecular and biochemical parasitology, 31(2), 127-131 (1988-11-01)
We have digested trypanosome DNA with a combination of pancreatic DNase I, nuclease P1 and bovine alkaline phosphatase and fractionated the resulting nucleosides on a Supelcosil LC-18-S column by high pressure liquid chromatography. We find less than 0.1% unusual nucleosides
Catrina L Dupard-Julien et al.
Analytical and bioanalytical chemistry, 387(3), 1027-1032 (2007-01-06)
A reversed-phase, high-performance liquid chromatography (RP-HPLC) method that allows quantitation of low levels of epoxides has been described. The method involved derivatization of epoxides using 100- to 1,000-fold excess N,N-diethyldithiocarbamate (DTC) at 60 degrees C for 20 min at neutral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service