Key Documents
57124
Supelco
Visidry Drying Attachment
for use with Visiprep 24-port model
About This Item
UNSPSC Code:
12000000
Pricing and availability is not currently available.
Agency
suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010
reg. compliance
suitable for FDA C-010.02
technique(s)
solid phase extraction (SPE): suitable
application(s)
food and beverages
compatibility
for use with Visiprep 24-port model
General description
The Visidry Drying Attachment was designed for our Visiprep Vacuum Manifold (Cat. No. 57100-U also fits our economical Preppy manifold). The Visidry unit installs in minutes, dries up 12 or up to 24 SPE tubes at one time, and can be used with any inert gas supply. It is also useful for evaporating and concentrating recovered samples. Gas flow to each port can be independently adjusted.
Note: The Visidry drying attachment cannot be used to dry 12mL, 20mL, or 60mL SPE tubes.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Related Content
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.
FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.