Skip to Content
Merck
All Photos(1)

Key Documents

16019AST

Supelco

Astec® CHIROBIOTIC® T2 Chiral (5 μm) HPLC Columns

L × I.D. 15 cm × 2.1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Product Name

Astec® CHIROBIOTIC® T2 Chiral HPLC Column, 5 μm particle size, L × I.D. 15 cm × 2.1 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L63

description

HPLC column

product line

Astec®

packaging

pkg of 1 ea

manufacturer/tradename

Astec®

parameter

0-45 °C temperature
241 bar pressure (3500 psi)

technique(s)

HPLC: suitable
LC/MS: suitable

L × I.D.

15 cm × 2.1 mm

matrix

high-purity silica gel particle platform
fully porous particle

particle size

5 μm

pore size

200 Å

operating pH range

3.8-6.8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

General description

CHIROBIOTIC® T and T2 have teicoplanin as the chiral selector. They offer unique selectivity for a number of classes of molecules, specifically underivatized α, β, γ and cyclic amino acids, N-derivatized amino acids, hydroxy-carboxylic acids, acidic compounds including carboxylic acids and phenols, small peptides, neutral aromatic analytes and cyclic aromatic and aliphatic amines. Separations normally obtained on a chiral crown ether or ligand exchange-type CSPs are also possible on the CHIROBIOTIC® T and T2, but in much simpler mobile phases, like water-alcohol. In addition, all of the known beta-blockers (amino alcohols), and dihydrocoumarins have been resolved. CHIROBIOTIC® T and T2 differ in their bonding chemistry and the pore size of the support particle, giving them different selectivity and preparative capacity.

  • Bonded phase: Teicoplanin
  • Operating pH range: 3.8 - 6.8
  • Particle diameter: 5, 10 or 16 μm
  • Pore size: 100 Å (CHIROBIOTIC® T) or 200 Å (CHIROBIOTIC® T2)

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographic Separations and Analysis: Macrocyclic Glycopeptide Chiral Stationary Phases
Berthod, A.
Comprehensive Chirality, 8, 227-262 (2012)
Chromatography Liquid Chiral Separations
Xiao, T.L., et al.
Encyclopedia of Separation Science, 1-15 (2000)
Ping Sun et al.
Journal of molecular structure, 890(1-3), 75-80 (2008-11-12)
A high performance liquid chromatographic method using macrocyclic glycopeptide chiral stationary phases (CSPs) was used to separate enantiomers of seven ruthenium(II) polypyridyl complexes. Among the five different CSPs, the Chirobiotic T2 was most effective and baseline separated all complexes. All
Chris Hamman et al.
Journal of chromatography. A, 1305, 310-319 (2013-07-31)
A rapid screening method to identify the best conditions for chiral separations is described. We analyzed a representative set of 80 racemic compounds against 25 different chiral stationary phases with three different mobile phases to identify the combination of columns
Sarangan Ravichandran et al.
Journal of chromatography. A, 1269, 218-225 (2012-08-25)
The enantioselective separations of the chiral oxazaphosphorines (R,S)-ifosfamide (IF), (R,S)-2-N-dechloroethyl-IF (2-DCE-IF) and (R,S)-3-N-dechloroethyl-IF (3-DCE-IF) were achieved on teicoplanin-based chiral stationary phase using isopropanol:methanol (60:40, v/v) as the mobile phase. Computational models of the teicoplanin and teicoplanin aglycon (TAG) chiral selectors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service