Z744105
CoolSafe™ system
to hold, 32 x 5 mL tubes
Synonym(s):
Cooling Box, cooling block, dry block, tube cooling block
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41123300
Recommended Products
feature
to hold 32 x 5 mL tubes
packaging
pack of 1 ea
manufacturer/tradename
Diversified Biotech CSAF-4000
Looking for similar products? Visit Product Comparison Guide
General description
System Includes: 1 CoolSafe Box with Lid, 1 CHAM-1000, One 1ºC and one -10ºC Cool Bricks. Versatile - Designed for 0.2ml tubes (strips or trays), 96-well and 384-well microplates, 0.5ml, 1.5ml, 5ml tubes. Specially Formulated - Cool Bricks maintain a constant temperature (1°C or –10°C) for up to 10 hours.
Application
PCR
Features and Benefits
Cool Bricks maintain a constant temperature (1°C or –10°C) for up to 10 hours.
Legal Information
CoolSafe is a trademark of Diversified Biotech, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service