Skip to Content
Merck
All Photos(1)

Key Documents

SRP5117

Sigma-Aldrich

PCNA, His tagged human

recombinant, expressed in E. coli, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

MGC8367

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.32

biological source

human

recombinant

expressed in E. coli

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

mol wt

~37 kDa

NCBI accession no.

application(s)

cell analysis

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... PCNA(5111)

General description

PCNA (proliferating cell nuclear antigen) gene is mapped to human chromosome 20p12.3 and encodes a scaffolding protein. PCNA is a homotrimeric protein known to localize in the nucleus. The PCNA ring has a central cavity that is large enough to allow the dsDNA to slide freely along it. PCNA is a nuclear protein whose appearance correlates with the proliferative state of the cells and is a cofactor of DNA polymerase delta.

Biochem/physiol Actions

Proliferating cell nuclear antigen (PCNA) significantly maintains the longevity of mature neutrophil granulocytes. Upregulation of the PCNA gene is observed in cervical squamous cell carcinoma. PCNA supports cell survival by elevating metabolism flux. PCNA expression is known to offer resistance to chemotherapy. PCNA is a homotrimer and helps increase the processivity of leading strand synthesis during DNA replication. In response to DNA damage, PCNA is ubiquitinated and is involved in the RAD6-dependent DNA repair pathway. Immunofluorescence studies have shown that p300 may play a role in DNA repair synthesis through its interaction with PCNA. In vitro and in vivo p300 forms a complex with PCNA that does not depend on the S phase of the cell cycle and stimulates DNA synthesis in vitro. PCNA interacts with the Williams syndrome transcription factor (WSTF) allowing it to target to DNA replication foci, that then allows recruitment of SNF2H.

Physical form

Supplied in 50mM sodium phosphate, pH 7.0, 300mM NaCl, 150mM imidazole, 0.1mM PMSF, 0.25mM DTT, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hisham Mazal et al.
eLife, 11 (2022-05-27)
Cryogenic optical localization in three dimensions (COLD) was recently shown to resolve up to four binding sites on a single protein. However, because COLD relies on intensity fluctuations that result from the blinking behavior of fluorophores, it is limited to
Proliferating cell nuclear antigen prevents trinucleotide repeat expansions by promoting repeat deletion and hairpin removal
Beaver JM, et al.
DNA Repair, 48(2), 17-29 (2016)
Cytoplasmic proliferating cell nuclear antigen connects glycolysis and cell survival in acute myeloid leukemia
Ohayon D, et al.
Scientific reports, 6(3), 35561-35561 (2016)
Replication Protein A Prohibits Diffusion of the PCNA Sliding Clamp along Single-Stranded DNA.
Hedglin M and Benkovic SJ
Biochemistry, 56(13), 1824-1835 (2017)
Raymond A Poot et al.
Nature cell biology, 6(12), 1236-1244 (2004-11-16)
Chromatin states have to be faithfully duplicated during DNA replication to maintain cell identity. It is unclear whether or how ATP-dependent chromatin-remodelling factors are involved in this process. Here we provide evidence that the Williams syndrome transcription factor (WSTF) is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service