SRP3008
APRIL human
recombinant, expressed in Hi-5 Insect cells, ≥95% (SDS-PAGE), ≥95% (HPLC), suitable for cell culture
Synonym(s):
A Proliferating-inducing Ligand, TNFSF13, TRDL-1?
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
biological source
human
recombinant
expressed in Hi-5 Insect cells
Assay
≥95% (HPLC)
≥95% (SDS-PAGE)
form
lyophilized
mol wt
16.3 kDa
packaging
pkg of 10 μg
technique(s)
cell culture | mammalian: suitable
impurities
<0.1 EU/μg endotoxin, tested
color
white to off-white
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... TNFSF13(8741)
General description
APRIL, a member of the TNF superfamily, is expressed in monocytes, macrophages, certain transformed cell lines, certain cancers of colon, and lymphoid tissues. APRIL, along with another TNF family member, BAFF, compete for two receptors, TACI and BCMA. ARPIL has the ability to stimulate proliferation of various tumor cell lines including Jurkat T cells and MCF-7 carcinoma cells. Like BAFF, APRIL also stimulates the proliferation of B and T cells. The human APRIL gene codes for at least four alternatively spliced transcriptional variants, which give rise to different isoforms of the APRIL precursor protein. All isoforms can be cleaved by the protease, furin, to release a soluble C-terminal fragment, which comprises the TNF like receptor binding of the APRIL precursor. Recombinant human APRIL is a soluble 16.3 kDa protein, consisting of 146 amino acid residues. 11
Biochem/physiol Actions
APRIL, a member of the TNF superfamily, is expressed in monocytes, macrophages, certain transformed cell lines, certain cancers of colon, and lymphoid tissues. Recombinant human APRIL is a soluble 16.3 kDa protein, consisting of 146 amino acid residues.
Sequence
AVLTQKQKKQ HSVLHLVPIN ATSKDDSDVT EVMWQPALRR GRGLQAQGYG VRIQDAGVYL LYSQVLFQDV TFTMGQVVSR EGQGRQETLF RCIRSMPSHP DRAYNSCYSA GVFHLHQGDI LSVIIPRARA KLNLSPHGTF LGFVKL
Physical form
Lyophilized from 20 mM Sodium Acetate, pH 7.5 + 500 mM NaCl.
Reconstitution
Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
G Yu et al.
Nature immunology, 1(3), 252-256 (2001-03-23)
We report that the tumor neurosis factor homolog APRIL (a proliferation-inducing ligand) stimulates in vitro proliferation of primary B and T cells and increases spleen weight due to accumulation of B cells in vivo. APRIL functions via binding to BCMA
Fabienne Mackay et al.
Cytokine & growth factor reviews, 14(3-4), 311-324 (2003-06-06)
B cell activating factor belonging to the TNF family (BAFF) and apoptosis-inducing ligand (APRIL) are two related members of the TNF ligand superfamily. Although they share two receptors, TACI and BCMA, transgenic and knockout mice in this system reveal that
H B Shu et al.
Journal of leukocyte biology, 65(5), 680-683 (1999-05-20)
Members of the tumor necrosis factor (TNF) family play important roles in modulation of immune responses. We describe the identification and cloning of a novel TNF family member that has been designated as TALL-1. TALL-1 is a 285-amino acid type
Yoshiko Matsuda et al.
Cellular immunology, 295(2), 127-136 (2015-04-17)
Long-lived plasma cells (PCs) contribute to humoral immunity through an undefined mechanism. Memory B cells, but not human naïve B cells, can be induced to differentiate into long-lived PCs in vitro. Because evidence links a proliferation-inducing ligand (APRIL), a tumor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service